Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Safety Tags

Catalog No. 17998145 Shop All AccuformNMC Products
Change view
Click to view available options
Message:
CAUTION
DANGER
DO NOT OPERATE
DO NOT START
DO NOT USE
Material:
Pulp-free Cardstock
RP Plastic
Length (Metric):
13 cm
14.6 cm
Style:
Standard back A
Standard back B
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Message Material Length (Metric) Style
17-998-145 DO NOT USE RP Plastic 13 cm Standard back B
17-998-137 DO NOT OPERATE RP Plastic 13 cm Standard back B
17-998-138 DO NOT START Pulp-free Cardstock 14.6 cm Standard back A
17-998-135 DO NOT OPERATE Pulp-free Cardstock 14.6 cm Standard back A
17-998-142 DO NOT USE Pulp-free Cardstock 14.6 cm Standard back A
17-998-143 DO NOT USE RP Plastic 13 cm Standard back A
17-998-139 DO NOT START RP Plastic 13 cm Standard back A
17-998-146 DANGER Pulp-free Cardstock 14.6 cm Standard back B
17-998-147 DANGER RP Plastic 13 cm Standard back B
17-998-136 DO NOT OPERATE Pulp-free Cardstock 14.6 cm Standard back B
17-998-144 DO NOT USE Pulp-free Cardstock 14.6 cm Standard back B
17-998-140 DO NOT START Pulp-free Cardstock 14.6 cm Standard back B
17-998-141 DO NOT START RP Plastic 13 cm Standard back B
17-998-148 CAUTION Pulp-free Cardstock 14.6 cm Standard back A
17-998-149 CAUTION RP Plastic 13 cm Standard back A
17-998-151 CAUTION RP Plastic 13 cm Standard back B
17-998-150 CAUTION Pulp-free Cardstock 14.6 cm Standard back B
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 17-998-145 Supplier Accuform Signs Supplier No. MDT217PTP
Add to Cart
Add to Cart

Provide immediate identification of the hazard by using the appropriate message for your tag

Specifications

Diameter (English) Grommet 0.37 in.
Material RP Plastic
Finish Matte
Message DO NOT USE
For Use With (Application) For indoor and outdoor conditions
Length (Metric) 13 cm
Length (English) 5.12 in.
Thickness 15 mil
Style Standard back B
Width (English) 3.3 in.
Width (Metric) 8.38 cm
Dimensions (L x W) 5.12 x 3.3 in. (13 x 8.38 cm)
Diameter (Metric) Hole 0.93 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.