Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Striped Marking Tapes

Catalog No. 19805176 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
Red
Yellow
Length (English):
54 ft.
108 ft.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Length (English)
19-805-176 Red 54 ft.
19-805-177 Yellow 108 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-805-176 Supplier Accuform Signs Supplier No. PTM734RDWT
Add to Cart
Add to Cart

Colored tapes are perfect for 5S programs and marking things in the work area.

Specifications

Adhesive Type Permanent Adhesive
Finish Overlaminate clear surface film
Width (English) 3 in.
Length (English) 54 ft.
Length (Metric) 16.46 m
Packaging Quantity 1 Roll
Certifications/Compliance OSHA 1910.144
For Use With (Application) Mark specific areas and promote safety by identifying potential hazard areas, marking fire protection and equipment per OSHA 1910.144
For Use With (Equipment) Fire Protection Equipment
Thickness 6 mil
Color Red
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.