Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform™ Lockout/Tagout Tags

Catalog No. 17998023 Shop All AccuformNMC Products
Change view
Click to view available options
Displayed Text:
Do Not Operate
Do Not Operate Equipment Lock Out
Do Not Operate Equipment Locked Out By
Do Not Operate Maintenance Department
Do Not Operate This Lock/Tag May Only be Removed By
Do Not Start
Equipment Locked Out
Locked Out Do Not Operate
Material:
HS-Laminate
PF Cardstock
RP Plastic
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Displayed Text Material
17-998-023 Do Not Operate Equipment Locked Out By RP Plastic
17-998-031 Do Not Operate Maintenance Department HS-Laminate
17-998-029 Do Not Operate Maintenance Department PF Cardstock
17-998-030 Do Not Operate Maintenance Department RP Plastic
17-998-025 Do Not Operate PF Cardstock
17-998-026 Do Not Operate RP Plastic
17-998-036 Do Not Start PF Cardstock
17-998-037 Do Not Start HS-Laminate
17-998-019 Do Not Operate Equipment Lock Out PF Cardstock
17-998-021 Do Not Operate Equipment Lock Out HS-Laminate
17-998-020 Do Not Operate Equipment Lock Out RP Plastic
17-998-027 Do Not Operate This Lock/Tag May Only be Removed By PF Cardstock
17-998-028 Do Not Operate This Lock/Tag May Only be Removed By RP Plastic
17-998-035 Locked Out Do Not Operate PF Cardstock
17-998-024 Do Not Operate Equipment Locked Out By HS-Laminate
17-998-022 Do Not Operate Equipment Locked Out By PF Cardstock
17-998-032 Equipment Locked Out PF Cardstock
17-998-034 Equipment Locked Out HS-Laminate
17-998-033 Equipment Locked Out RP Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Catalog No. 17-998-023 Supplier Accuform™ Supplier No. MLT409PTP
Add to Cart
Add to Cart

Provide immediate identification of the hazard by using the appropriate message for your tag.

Specifications

Product Type Lockout/Tagout Tag
Material RP Plastic
Certifications/Compliance OSHA 29 CFR 1910.147
Dimensions (L x W) 5.12 x 3.3 in. (130 x 84 mm)
Displayed Text Do Not Operate Equipment Locked Out By
For Use With (Application) Identify clearly and alert staff to cautions with durable safety tags that work effectively in extreme environments
Length (English) 5.12 in.
Length (Metric) 130 mm
Thickness 15 mil
Width (English) 3.3 in.
Width (Metric) 84 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.