Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Voltage Markers

Catalog No. 19804574 Shop All AccuformNMC Products
Change view
Click to view available options
Quantity:
1 Ea.
4/Pk.
Displayed Text:
120 Volts
208 Volts
240 Volts
440 Volts
480 Volts
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Displayed Text
19-804-574 1 Ea. 440 Volts
19-804-571 1 Ea. 120 Volts
19-804-573 1 Ea. 240 Volts
19-804-572 4/Pk. 208 Volts
19-821-785 4/Pk. 480 Volts
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-804-574 Supplier Accuform Signs Supplier No. LELC434
Add to Cart
Add to Cart

Identify high voltage and electrical hazards with Conduit Voltage Markers

Specifications

Material Vinyl
Quantity 1 Ea.
Size (English) 1.12 x 4.5 in.
Size (Metric) 2.84 x 11.4 cm
Mounting Type Self-adhesive
For Use With (Application) The durable adhesive will stick to most flat, clean surfaces, Adhesive Vinyl signs will work for short term outdoor applications and in moderate temperature
Thickness 4 mil
Temperature Range (Metric) -20°C to +80°C
Temperature Range (English) -20°F to +176°F
Displayed Text 440 Volts
Product Type Voltage Marker
Life Span 2 to 4 Years
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.