Learn More
Promega AccuMAP 100X Oxidation Suppressant, 50microl
Catalog No. PRVA1020 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
50 μL
Catalog No. Quantity
PRVA1020 50 μL
1 options
Catalog No. PRVA1020 Supplier Promega Supplier No. VA1020
AccuMAP Low pH Protein Digestion Kit enables suppression of sample preparation-induced artificial deamidation, disulfide bond scrambling and oxidation overthe course of sample preparation. Pre-existing non-enzymatic modifications remain intact.
  • Suppress Artificial Deamidation and Disulfide Bond Scrambling in Protein Sample Prep
  • Sample preparation completed in 4.5−5 hours
  • Highly reproducible digestion results
  • Use for reduced and nonreduced proteins
Specifications
Content And Storage <-65°C
Quantity 50 μL