Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ accumet™ AB150 Benchtop pH Meters

Catalog No. 13636AB150 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Meter with BioBasic Kit
Meter with BioBasic Kit, NIST
Meter with pH Kit
Meter with pH Kit, NIST Cert.
pH Meter
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13-636-AB150 Meter with pH Kit
13-636-AB150B Meter with BioBasic Kit
13-636-AB150A pH Meter
AB150BCERT Meter with BioBasic Kit, NIST
AB150CERT Meter with pH Kit, NIST Cert.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 13-636-AB150
This item is not returnable. View return policy

Includes

accumet AB150 pH benchtop meter, TRIS-compatible double junction refillable epoxy-body pH/ATC electrode (13-620-631), electrode stand, RS232 and USB cables, 100/240V universal power supply

Provide intuitive, simple operation and high accuracy measurements in a compact, affordable meter. Fisherbrand accumet AB150 pH benchtop meters have large backlit display which offers great visibility in laboratory environments. The accumet AB150 pH meter is an industry favorite for when reliability and consistency in measurements matter.

  • Graphic backlit display with date and time, stability indicator and alarms
  • One to six point pH calibration with 0.001 pH resolution
  • Automatic buffer recognition with four pH buffer sets or custom buffer option
  • pH slope and offset display
  • One point relative mV calibration
  • One point ATC temperature probe calibration
  • Automatic or manual temperature compensation, displayed in °C or °F
  • Password protection of calibration and setup modes
  • 500 data set storage
  • RS232 and USB interfaces
  • Multilanguage interface with English, German, Chinese, French, Italian and Spanish
  • Optional stirring probe (Cat. No. 13-620-BSP) with five speed settings

NIST-Traceable Calibration Models (-CERT)

  • Help meet ISO, FDA, USDA and EPA quality assurance guidelines
  • Includes meter accuracy at five test points across pH range
  • Includes meter accuracy at five test points across corresponding mV range
  • Testing performed by accredited laboratory; includes NIST-Traceable certificate

Specifications

  • pH Range: -2.000 to 20.000 pH
  • pH Resolution: 0.1, 0.01, 0.001 pH
  • pH Accuracy: ±0.002 pH
  • mV Range: ±2000.0mV
  • mV Resolution: 0.1mV
  • mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
  • mV Offset Adjustment: Up to ±150mV
  • Temperature Range: 0° to 100°C (32° to 212.0°F)
  • Temperature Resolution: 0.1°C (0.1°F)
  • Temperature Accuracy: ±0.3°C (±0.5°F)
  • Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
  • ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
  • Self-Test: Yes
  • Inputs: BNC, mini-phone ATC, ground
  • Power Requirements: 100 to 240VAC, 50/60Hz

Applications
Academic, research, QA/QC, drinking water, wastewater, food, beverage, industrial

Certifications

Select models available with Certificate of NIST Traceability

Warranty and Services

3 years on meter

TRUSTED_SUSTAINABILITY

Specifications

Accuracy (mV) ±0.2
Data Management Details Manual, Timed (selectable every 3 to 3600 sec.), Printer or CSV Format
Accuracy (Temperature) ±0.3°C
Accuracy (pH) ±0.002
Outputs RS-232 (phono plug), mini-B USB, stirrer
Includes Meter, TRIS Compatible pH/ATC Electrode (13-620-631), Electrode Arm, RS-232 and USB Cables, 100/240V Power Supply
Range (pH) -2 to 20
Resolution (pH) 0.001, 0.01, 0.1
Calibration Points (pH) Up to 6 pre-set or 5 custom
Range (mV) ±2000
Resolution (mV) 0.1 mV
Calibration Points 1 to 6
Temperature Range (Metric) 0°C to 100°C
Resolution (Temperature) 0.1°C
Display Type LCD
Data Points 500
Inputs BNC, ATC
Certifications/Compliance Without NIST-traceable pH Certificate
Warranty 3 Years
Model AB150 pH Kit
Type Meter with pH Kit
Voltage 100/240 V
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.