Learn More
Description
Includes
accumet AB150 pH benchtop meter, electrode stand, RS232 and USB cables, 100/240V universal power supply
Provide intuitive, simple operation and high accuracy measurements in a compact, affordable meter. Fisherbrand accumet AB150 pH benchtop meters have large backlit display which offers great visibility in laboratory environments. The accumet AB150 pH meter is an industry favorite for when reliability and consistency in measurements matter.
- Graphic backlit display with date and time, stability indicator and alarms
- One to six point pH calibration with 0.001 pH resolution
- Automatic buffer recognition with four pH buffer sets or custom buffer option
- pH slope and offset display
- One point relative mV calibration
- One point ATC temperature probe calibration
- Automatic or manual temperature compensation, displayed in °C or °F
- Password protection of calibration and setup modes
- 500 data set storage
- RS232 and USB interfaces
- Multilanguage interface with English, German, Chinese, French, Italian and Spanish
- Optional stirring probe (Cat. No. 13-620-BSP) with five speed settings
NIST-Traceable Calibration Models (-CERT)
- Help meet ISO, FDA, USDA and EPA quality assurance guidelines
- Includes meter accuracy at five test points across pH range
- Includes meter accuracy at five test points across corresponding mV range
- Testing performed by accredited laboratory; includes NIST-Traceable certificate
Specifications
- pH Range: -2.000 to 20.000 pH
- pH Resolution: 0.1, 0.01, 0.001 pH
- pH Accuracy: ±0.002 pH
- mV Range: ±2000.0mV
- mV Resolution: 0.1mV
- mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
- mV Offset Adjustment: Up to ±150mV
- Temperature Range: 0° to 100°C (32° to 212.0°F)
- Temperature Resolution: 0.1°C (0.1°F)
- Temperature Accuracy: ±0.3°C (±0.5°F)
- Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
- ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
- Self-Test: Yes
- Inputs: BNC, mini-phone ATC, ground
- Power Requirements: 100 to 240VAC, 50/60Hz
Applications
Academic, research, QA/QC, drinking water, wastewater, food, beverage, industrial
Certifications
Select models available with Certificate of NIST Traceability
Warranty and Services
3 years on meter
Specifications
Specifications
| Accuracy (Temperature) | ±0.3°C |
| Data Management Details | Manual, Timed (selectable every 3 to 3600 sec.), Printer or CSV Format |
| Accuracy (mV) | ±0.2 |
| Accuracy (pH) | ±0.002 |
| Outputs | RS-232 (phono plug), mini-B USB, stirrer |
| Includes | Meter, Electrode Arm, RS-232 and USB Cables, 100/240V Power Supply |
| Range (pH) | -2 to 20 |
| Resolution (pH) | 0.001, 0.01, 0.1 |
| Calibration Points (pH) | Up to 6 pre-set or 5 custom |
| Range (mV) | ±2000 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.