Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ accumet™ AE150 Benchtop pH Meter

Catalog No. p-8557008 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
3-in-1 Set
Bio Set
Education Set
Standard Set
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13-636-AE150 Standard Set
13-636-AE151 Education Set
13-636-AE152 Bio Set
13-636-AE153 3-in-1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13-636-AE150

Includes

Meter, Electrode Stand, Univ. Power Adapter. (pH Probe and Temperature Probe sold separately.)

Provides the essential functions needed for routine pH measurements without complicated features and processes at a budget-friendly price

Designed for routine pH measurements in the lab with convenience in mind – simple functionality, stored data sets in a non-volatile memory and meter-attachable electrode stand. The accumet AE150 pH meters offer efficient sample reading and recording with flexible display options to show display both pH and temperature or mV and temperature simultaneously.

  • One to five point pH calibration
  • Automatic buffer recognition with two pH buffer sets
  • pH slope and offset display
  • One point mV calibration
  • One point ATC temperature probe calibration
  • Automatic or manual temperature compensation
  • Ready icon to indicate measurement stability
  • Electrode condition icon to indicate status
  • 100 data set storage with non-volatile memory
  • Note: pH and/or temperature probe is not included

Warranty and Services

3 years on meter

TRUSTED_SUSTAINABILITY

Specifications

Accuracy (Temperature) ±0.3°C
Accuracy (mV) ±0.2 ±199.9 ±2 beyond ±200
Accuracy (pH) ±0.01
Outputs Recorder (±2000 mV)
Includes Meter, Electrode Stand, Univ. Power Adapter (90-240V, 50/60 Hz)
Calibration USA: pH 1.68, 4.01, 7.00, 10.01, 12.45
Range (pH) 0 to 14
Range (mV) ±1999
Resolution (mV) 0.1 mV (±199.9 mV); 1 mV beyond ±200 mV
Calibration Points 1 to 5 with automatic buffer recognition (ABR)
Temperature Range (Metric) 0.0°C to 100.0°C
Resolution (Temperature) 0.1°C
Temperature Selection Automatic or manual (0 to 100 °C)
Temperature Probe Calibration Offset in 0.1 °C increments; Offset range: ±5 °C
Memory 100 data sets
Inputs BNC, 2.5 mm phono plug, reference pin tip, ground
Warranty 3 Year Meter Warranty
Probe Type Probe is not included (sold separately).
Range (Relative mV) Up to ±150 mV
Type Standard Set
Electrical Requirements AC/DC 9 V adapter (110/220 VAC, 50-60 Hz)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.