Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ accumet™ Four Cell Conductivity/ATC Probes, Four-Cell

Catalog No. 13620165 Shop All Fisherbrand Products
Change view
Click to view available options
Cell Constant:
1 cm-1
10 cm-1
Measurement Ranges:
1 to 200 mS/cm
10 μS to 100 mS/cm
Material:
Epoxy
Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cell Constant Measurement Ranges Material
13-620-165 1 cm-1 10 μS to 100 mS/cm Epoxy
13-620-166 10 cm-1 1 to 200 mS/cm Epoxy
13-620-163 1 cm-1 10 μS to 100 mS/cm Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-620-165
Add to Cart
Add to Cart

For use with AR60, AR50, AR20, AB30 benchtop and AP65 portable meters

Immersion-type cell's unique compact size is the same as a typical pH electrode—allowing use in standard electrode holder.

  • Four-cell type with glass or epoxy bodies
  • Built-in temperature sensors and 8-pin connectors
  • Cell constants of 0.1, 1.0, or 10.0cm-1

TRUSTED_SUSTAINABILITY

Specifications

Cell Constant 1 cm-1
Type Conductivity/ATC Probe
For Use With (Equipment) Fisher Scientific accumet benchtop models XL600, XL500, XL200, & AB200 and legacy models XL60, XL50, XL30, XL20, AR60, AR50, AR30, AR20, & AB30
Connector Types 8-pin
Material Epoxy
Measurement Ranges 10 μS to 100 mS/cm
Storage Method Immersed in deionized water
Probe 4 Cell Conductivity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.