Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ accumet™ XL200 Benchtop pH/Conductivity Meters

Catalog No. XL200CERT Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Meter with pH/Cond. Kit
Meter, pH/Cond. Kit, NIST Cert.
pH/Cond Meter, NIST Cert.
pH/Conductivity Meter
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
XL200CERT pH/Conductivity Meter
13-636-XL200 pH/Cond Meter, NIST Cert.
13-636-XL200A Meter, pH/Cond. Kit, NIST Cert.
XL200ACERT Meter with pH/Cond. Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. XL200CERT

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Includes

accumet XL200 pH/conductivity benchtop meter, TRIS-compatible double junction refillable glass-body pH electrode (13-620-183A), ATC probe (13-620-19), 2-cell conductivity/temperature probe (13-620-100), electrode stand, RS232 and USB cables, 100/240V universal power supply, NIST traceability certificate

Provide advanced measurements with simple operation and offer a large, vivid color touch-screen display that allows for dynamic functionality. Fisherbrand accumet XL200 pH/Conductivity benchtop meters offer advanced features with simple operation to help ensure your measurements are reliable and your laboratory processes are efficient.

  • High resolution color display shows data clearly from wide viewing angles and distances
  • Optimized touch-screen with glass/glass sensitivity provides great ease of use
  • Onscreen info includes date and time, stability indicator, alarms and context-specific help screens
  • One to six point pH calibration with 0.001 pH resolution
  • Automatic buffer recognition with six pH buffer sets or custom buffer option
  • Conductivity, TDS (total dissolved solids), salinity and resistivity modes
  • Automatic or manual temperature compensation, displayed in °C or °F
  • Display up to three measurements simultaneously (two pH or ISE, one conductivity)
  • Up to 10 password protected user ID accounts, each retains unique calibration and instrument settings with memory to store 2000 data points
  • RS232 and USB interfaces for printer or computer interfacing and real-time onscreen graphing of measurements
  • RJ45 Ethernet/Internet port to connect Local Area Network (LAN) or Internet
  • Bidirectional communication for LIMS connectivity and 3rd party software
  • Connectivity for peripherals such as keyboard, mouse or other USB device
  • Multilanguage interface with English, German, Chinese, French, Italian and Spanish
  • Two ports for optional stirring probe (Cat. No. 13-620-BSP) with five speed settings
  • Option to wall-mount meter to create additional bench space

NIST-Traceable Calibration Models (-CERT)

  • Help meet ISO, FDA, USDA and EPA quality assurance guidelines
  • Includes meter accuracy at five test points across pH range
  • Includes meter accuracy at five test points across corresponding mV range
  • Testing performed by accredited laboratory, includes NIST-Traceable certificate

Specifications

  • pH Range: -2.000 to 20.000 pH
  • pH Resolution: 0.1, 0.01, 0.001 pH
  • pH Accuracy: ±0.002 pH
  • ISE Range: 0.001 to 19999 ppm, mg/L or molar
  • ISE Resolution: 0.001, 0.01, 0.1, 1 (automatic)
  • ISE Accuracy: 0.5 % full scale (monovalent ion); 1 % full scale (divalent ion)
  • mV Range: ±2000.0mV
  • mV Resolution: 0.1mV
  • mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
  • mV Offset Adjustment: Up to ±150mV
  • Conductivity Range: 0.00μS to 500.0mS
  • Conductivity Resolution: 0.01, 0.1μS; 0.001, 0.01, 0.1mS
  • Conductivity Accuracy: ±1 % full scale
  • TDS Range: 0.00ppm to 500 ppt (TDS factor 1.00)
  • TDS Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
  • TDS Accuracy: ±1 % full scale
  • Salinity Range: 0 to 80.0 ppt
  • Salinity Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
  • Salinity Accuracy: ±1 % full scale
  • Resistivity Range: 2.000 Ohm to 20.0 MOhm
  • Resistivity Resolution: 0.01, 0.1 Ohm; 0.001, 0.1 kOhm; 0.01 MOhm
  • Resistivity Accuracy: ±1 % full scale
  • Temperature Range: -5° to +105°C (23° to 221.0°F)
  • Temperature Resolution: 0.1°C (0.1°F)
  • Temperature Accuracy: ±0.2°C (±0.3°F)
  • Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
  • ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
  • Memory: 2000 data sets per user ID with date/time stamp
  • Self-Test: Yes
  • Inputs: BNC, mini-phone ATC, 8 Pin DIN (conductivity), ground
  • Power Requirements: 100 to 240VAC, 50/60Hz
  • Dimensions (L x W x H): 24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.)
  • Weight: 1.13kg (2.5 lb.)

Applications
Pharmaceutical, biotech, research, QA/QC, industrial, food, beverage

Certifications

CE, TUV 3-1, FCC Class A

Warranty and Services

3 years on meter

TRUSTED_SUSTAINABILITY

Specifications

Outputs RS232, USB-A, Mini-B USB, RJ45, stirrer port (2)
Accuracy Multiple
Accuracy (Conductivity) ±1% FS
Accuracy (mV) ±0.2 mV
Accuracy (pH) ±0.002
Accuracy (Resistivity) ± 1% full scale
Accuracy (Salinity) ± 1% full scale
Accuracy (TDS) ± 1% full scale
Calibration Points (Conductivity) Automatic (4 points), maximum 1/range, manual (5 points), maximum 1/range
Calibration Points (pH) Up to 6 preset or 5 custom
Cell Constant 0.01 to 10
Certifications/Compliance NIST, ISO, FDA, USDA, EPA
Data Management Details 3 to 86400 sec.
Data Points 2000
Detection Range Multiple
Display Type LCD
Range (Conductivity) 0 to 500 mS
Range (mV) ±2000 mV
Range (pH) -2 to 20
Range (Resistivity) 3 W to 100 MW over 4 ranges: 0 to 20 kW, 20 to 200 kW, 200 kW to 2 MW, 2 to 100 MW
Range (Salinity) 0 to 80 ppt
Range (TDS) 0.050 ppm to 500 ppt (at TDS factor 1.00)
Resolution (pH) 0.1/0.01/0.001
Includes Meter, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), Conductivity/Temp probe (13-620-100), electrode arm, RS-232 and USB cables, 110/220 V power supply, and NIST-traceable certificate
Inputs BNC, ATC
Model XL200 pH/Conductivity Kit w/ NIST-Traceable Certificate
Product Type pH/Conductivity Meter
Resolution Multiple
Resolution (Conductivity) 0.001/0.01/0.1mS
Resolution (mV) 0.1mV
Resolution (Resistivity) 0.01/0.1W, 0.001/0.1kW, 0.01MW
Resolution (Salinity) 0.01/0.1ppm, 0.001/0.01/0.1ppt
Resolution (TDS) 0.01/0.1ppm, 0.001/0.01/0.1ppt
Temperature Accuracy ±0.2°C
Temperature Resolution 0.1°C
Warranty 3 Years
Dimensions (L x W x H) 24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.)
Length (English) 9.68 in.
Length (Metric) 24.6 cm
Width (Metric) 17.3 cm
Height (English) 3.89 in.
Height (Metric) 9.9 cm
Weight (English) 2.5 lb.
Weight (Metric) 1.13 kg
Voltage 100/240 V
Frequency 50/60Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.