Specifications
| Lens Coating | Anti-fog |
| Certifications/Compliance | ANSI Z87.1,CSA Z94.3-07 |
| Description | Full eye protection 10 curve lens |
| Lens Tint | Gray |
| Frame Color | Black |
| Temple Style | Spatula with double injection rubber |
| Lens Material | Polycarbonate |
| Frame Material | Nylon |
| Frame Type | Dual Lens with Side Protection |