Learn More
Description
- Manufactured from human serum or plasma
Specifications
Specifications
| Certifications/Compliance | IVD, EU, CE |
| Content And Storage | 2°C to 8°C |
| Format | Vial |
| Type | ACCURUN 113 HBc IgM Positive Control |
| Intended Use | Detection of IgM Antibodies to Hepatitis B core Antigen (HBc IgM) |
| Quantity | 1 x 5 mL |
| Source | Human Serum or Plasma |
| Stability | 60 Days Open Vial |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.