Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ ACCURUN™ QC Hepatitis Positive Controls

Catalog No. 22554171 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Type:
ACCURUN 113 HBc IgM Positive Control
ACCURUN 121 Anti-HAV IgM Positive Control
ACCURUN 125 anti-HBs Positive Control
Intended Use:
Detection of IgM Antibodies to Hepatitis B core Antigen (HBc IgM)
Qualitative detection of Antibodies to Hepatitis B Surface Antigen (Anti-HBs)
Qualitative determination of IgM Antibodies to Hepatitis A Virus (HAV IgM)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Intended Use
22-554-171 ACCURUN 121 Anti-HAV IgM Positive Control Qualitative determination of IgM Antibodies to Hepatitis A Virus (HAV IgM)
22-554-173 ACCURUN 125 anti-HBs Positive Control Qualitative detection of Antibodies to Hepatitis B Surface Antigen (Anti-HBs)
22-554-165 ACCURUN 113 HBc IgM Positive Control Detection of IgM Antibodies to Hepatitis B core Antigen (HBc IgM)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-554-171 Supplier SeraCare™ Supplier No. 20150074
Add to Cart
Add to Cart

Offer the ultimate in flexibility and convenience

  • Manufactured from human serum or plasma

Specifications

Certifications/Compliance IVD, EU, CE
Content And Storage 2°C to 8°C
Format Vial
Type ACCURUN 121 Anti-HAV IgM Positive Control
Intended Use Qualitative determination of IgM Antibodies to Hepatitis A Virus (HAV IgM)
Quantity 1 x 5 mL
Source Human Serum or Plasma
Stability 60 Days Open Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.