Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ ACCURUN™ QC Serology Multi-Marker Negative Controls

Catalog No. 22554127 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
1 x 1 mL
6 x 3.5 mL
Type:
ACCURUN 800 ToRCH Serology Negative Control
ACCURUN 810 Multi-analyte Serology Negative Control
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
22-554-127 1 x 1 mL ACCURUN 800 ToRCH Serology Negative Control
22-554-131 6 x 3.5 mL ACCURUN 810 Multi-analyte Serology Negative Control
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-554-127 Supplier SeraCare™ Supplier No. 20100018
Add to Cart
Add to Cart

Simplify testing by using one control vial for analytes commonly grouped in one laboratory area

  • Enhance quality control and quality assurance in your testing lab
  • Human plasma-based controls mimic patient samples
  • Independent controls meet compliance requirements of regulatory agencies and professional organizations
  • Liquid, ready-to-use–no reconstitution or special handling required
  • Extended shelf life means efficiency for your lab, allows you to monitor assay performance with same lot over an extended period of time

  • Combine LabLink xL with the ACCURUN products for an IVD exclusive infectious disease and nucleic acid testing Quality Assurance Program (QAP) from Fisher HealthCare
  • With the LabLink xL QAP program, a lab's performance can be compared to peer laboratories' performances and used to benchmark performance to group standards:
    • Track how other lab's signal to cutoff ratio (s/co) or recovered values (mean values) compare
    • Compare how a lab's variability (standard deviation) compares to a group average
    • Determine if a lab's s/co or recovered values are statistically within acceptable group limits

Compliance

For in vitro diagnostic use; CE-approved

Specifications

Certifications/Compliance IVD, CE, HC
Content And Storage 2°C to 8°C
For Use With (Equipment) Any system
Type ACCURUN 800 ToRCH Serology Negative Control
Intended Use Detection of IgG and IgM Antibodies to Toxoplasma gondii, Rubella Virus, Cytomegalovirus, and Herpes Simplex Virus
Quantity 1 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.