Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ ACCURUN™ QC Serology Multi-Marker Positive Controls

Catalog No. 22554119 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
1 x 5 mL
3 x 1 mL
6 x 3.5 mL
Type:
ACCURUN 21 1000 Multi-analyte Positive Control
ACCURUN 21 2000 Multi-analyte Positive Control
ACCURUN 25 ToRCH IgG Positive Control
ACCURUN 30 EBV IgG Positive Control
ACCURUN 31 EBV IgM Positive Control
ACCURUN 40 Multi-analyte Positive Control
ACCURUN 51 Multi-analyte Hepatitis Positive Control 1
ACCURUN 52 Multi-analyte Hepatitis Positive Control
ACCURUN ToRC IgM Positive Control
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
22-554-119 1 x 5 mL ACCURUN 52 Multi-analyte Hepatitis Positive Control
22-554-117 1 x 5 mL ACCURUN 51 Multi-analyte Hepatitis Positive Control 1
22-554-191 3 x 1 mL ACCURUN 40 Multi-analyte Positive Control
22-554-192 6 x 3.5 mL ACCURUN 21 1000 Multi-analyte Positive Control
22-554-124 3 x 1 mL ACCURUN 31 EBV IgM Positive Control
22-554-122 3 x 1 mL ACCURUN 25 ToRCH IgG Positive Control
22-554-120 6 x 3.5 mL ACCURUN 52 Multi-analyte Hepatitis Positive Control
22-554-123 3 x 1 mL ACCURUN 30 EBV IgG Positive Control
22-554-193 6 x 3.5 mL ACCURUN 21 2000 Multi-analyte Positive Control
22-554-118 6 x 3.5 mL ACCURUN 51 Multi-analyte Hepatitis Positive Control 1
22-554-190 3 x 1 mL ACCURUN ToRC IgM Positive Control
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 22-554-119 Supplier SeraCare™ Supplier No. A525001

Simplify testing by using one control vial for analytes commonly grouped in one laboratory area

  • Quality control for infectious disease testing laboratories
  • Detect and prevent errors
  • Monitor assay performance
  • Simplify testing procedures
  • Document regulatory compliance
  • Designed to mimic a patient sample
  • Supplied ready to use, requiring no reconstitution or special handling

Specifications

Content And Storage -20°C
Type ACCURUN 52 Multi-analyte Hepatitis Positive Control
Intended Use Qualitative determination of Hepatitis B e Antigen (HBeAg), IgM Antibodies to Hepatitis B core Antigen (HBc IgM), IgM Antibodies to Hepatitis A Virus (HAV IgM)
Quantity 1 x 5 mL
Source Human Serum or Plasma
Stability 60 Days Open Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.