Learn More
SeraCare™ ACCURUN™ QC Serology Multi-Marker Positive Controls
Shop All SeraCare Life Sciences ProductsDescription
- Quality control for infectious disease testing laboratories
- Detect and prevent errors
- Monitor assay performance
- Simplify testing procedures
- Document regulatory compliance
- Designed to mimic a patient sample
- Supplied ready to use, requiring no reconstitution or special handling
Specifications
Specifications
| Certifications/Compliance | ISO 13485, CE, IVD, HC |
| Content And Storage | 2°C to 8°C |
| Type | ACCURUN 40 Multi-analyte Positive Control |
| Intended Use | Qualitative determination of Antibodies to Measles, Mumps, Rubella, and VZV |
| Quantity | 3 x 1 mL |
| Source | Human Serum or Plasma |
| Stability | 60 Days Open Vial |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.