Learn More
SeraCare™ ACCURUN™ QC Serology Single Analyte Positive Controls
Shop All SeraCare Life Sciences ProductsDescription
Quality control for infectious disease testing laboratories.
- Detect and prevent errors
- Monitor assay performance
- Simplify testing procedures
- Document regulatory compliance
- Contain a single antibody or antigen and are designed to perform in the dynamic range of most manufacturers' assays
- Designed to mimic a patient sample
- Supplied ready to use, requiring no reconstitution or special handling
Specifications
Specifications
| Certifications/Compliance | IVD, CE |
| Content And Storage | -20°C |
| Format | Vial |
| For Use With (Equipment) | BioMerieux™ Vidas™ |
| Type | ACCURUN 132 Lyme IgM Positive Control |
| Intended Use | Qualitative detection of IgG Antibodies to Borrelia burgdorferi (Lyme disease) |
| Quantity | 1 x 1 mL |
| Source | Human Serum or Plasma |
| Stability | 60 Days Open Vial |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.