Learn More
World Precision Instrument Accuscope Trinocular Microscope, Plan phase contrast
SDP
Supplier:  World Precision Instrument 504444
Encompass
The ACCU-SCOPE 3000-LED Microscope Series delivers outstanding optical performance, value and resolution to meet the exacting standards of life science professionals and students. With a newly designed infinity plan optical system, a best-in-class 20 mm field of view and a super-bright 3-watt LED illuminator, the 3000-LED Series provides high contrast images with outstanding resolution, precision design and enhanced illumination.
Catalog No. 50-253-9374