Learn More
SeraCare™ AccuSpan™ Linearity Panels
Catalog No. 22554024 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Type:
Epstein Barr Virus (EBV)
Herpes Simplex Virus 1 (HSV1)
Herpes Simplex Virus 2 (HSV2)
JC Virus (JCV)
Varicella Zoster Virus (VZV)
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-554-024 Epstein Barr Virus (EBV)
22-554-028 Varicella Zoster Virus (VZV)
22-554-027 JC Virus (JCV)
22-554-026 Herpes Simplex Virus 2 (HSV2)
22-554-025 Herpes Simplex Virus 1 (HSV1)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-554-024 Supplier SeraCare™ Supplier No. 24100255
Add to Cart
Add to Cart
This item is not returnable. View return policy
For evaluating and monitoring molecular diagnostic assay procedures used for detection of viral nucleic acids
  • Each panel consists of four members: one negative, one low positive, one medium positive and one high positive sample
  • Comes in ready-to-go, single-use format with target concentrations that cover range of most molecular assays
  • Full size: 4 x 1mL
Specifications
Type Epstein Barr Virus (EBV)
Includes 1 negative, 1 low positive, 1 medium positive, 1 high positive sample
Quantity 4 x 1 mL