Learn More
Description
- Each panel consists of four members: one negative, one low positive, one medium positive and one high positive sample
- Comes in ready-to-go, single-use format with target concentrations that cover range of most molecular assays
- Full size: 4 x 1mL
Specifications
Specifications
| Type | Herpes Simplex Virus 2 (HSV2) |
| Includes | 1 negative, 1 low positive, 1 medium positive, 1 high positive sample |
| Quantity | 4 x 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.