Learn More
Bioneer AccuTarget™ Fluorescein-labeled miRNA Negative Control #1, HPLC, 5 nmol
SDP
Supplier:  Bioneer SMC4001
Encompass
Provides AccuTarget ™ miRNA mimic control for optimizing cell experimental conditions for miRNA mimic function studies. miRNA positive and negative controls can be used as controls in miRNA functional studies using our AccuTarget ™ miRNA mimic.Ready-to-transfect miRNA mimicSame activity as the endogenous miRNA actually present in the cell after transfectionWide ApplicationsmiRNA inhibitors for studying the loss-of-function of miRNAs by inhibiting the activity of target miRNAs
Catalog No. NC2751329