Learn More
LAXCO Accuva LEAP to complete LCM capability upgrade kit
SDP
Catalog No. 12007190 Shop All Laxco Inc Products
Change view
Click to view available options
Voltage:
110/220 V
Catalog No. Voltage
12-007-190 110/220 V
1 options
Catalog No. 12-007-190 Supplier LAXCO Supplier No. SS9LCMK3
Add to Cart
Add to Cart
Request a Quote
Full capability of Laser Capture Microdissection.
  • Upgrade Kit to enhance the Accuva LEAP system with full Laser Capture Microdissection (LCM) capability
  • Includes both hardware and software upgrades
  • Installation to be performed at manufacturer site
  • System will be sent back after installation is completed
  • Includes UV laser for cutting and microdissection
  • IR laser for cell capture
Specifications
Type LCM Upgrade Kit
Voltage 110/220 V