Learn More
LAXCO Accuva LEAP to laser cutting capability upgrade kit
SDP
Catalog No. 12007194 Shop All Laxco Inc Products
Change view
Click to view available options
Voltage:
110/220 V
Catalog No. Voltage
12-007-194 110/220 V
1 options
Catalog No. 12-007-194 Supplier LAXCO Supplier No. SS9UVK2
Add to Cart
Add to Cart
Request a Quote
Allows laser cutting on thin samples using UV laser.
  • LAXCO Accuva LEAP Laser Cutting Capability Upgrade Kit
  • Cell cutting laser damage and recovery
  • Enables laser cutting on thin samples using a UV laser
  • Mark and cut specific areas of interest for downstream analysis
  • Allows cutting of organelles and single cells (10 to15 μm)
  • Instrument to be shipped to LAXCO for upgrade and returned afterward
Specifications
Type Laser Cutting Upgrade Kit
Voltage 110/220 V