Learn More
LAXCO Accuva LEAP phase to multichannel fluorescence upgrade kit
SDP
Catalog No. 12007188 Shop All Laxco Inc Products
Change view
Click to view available options
Voltage:
110/220 V
Catalog No. Voltage
12-007-188 110/220 V
1 options
Catalog No. 12-007-188 Supplier LAXCO Supplier No. SS9FLK1
Add to Cart
Add to Cart
Request a Quote
Unique design allowing seamless integration.
  • LAXCO Accuva LEAP phase-to-multichannel fluorescence upgrade kit
  • Includes fluorescent LEDs and filter cubes
  • Enables the system to perform red, blue, and green (R/B/G) fluorescence
  • Professional installation included
  • Instrument shipped to LAXCO for upgrade and returned post-installation
Specifications
Type LED and Filter Cubes
Voltage 110/220 V