Learn More
LAXCO Accuva LEAP Upgradable Phase Inverted Imaging System PROMO
SDP
Catalog No. 12007191PM Shop All Laxco Inc Products
Change view
Click to view available options
Voltage:
110/220 V
Catalog No. Voltage
12-007-191PM 110/220 V
1 options
Catalog No. 12-007-191PM Supplier LAXCO Supplier No. SS9PH1PROMO26
Get SmartSTART & Leap AI Software with Purchase
View all promotions
Add to Cart
Add to Cart
Motorized X-Y-Z stage. Eyepieceless design.
  • Motorized 6-position objective turret with objectives: 2x, 4x, 10x, 20x, 40x, 60x Plan Phase LWD
  • Motorized Z-Axis with memory positions
  • Future-proof design with upgrade options to expand to fluorescent and LCM capabilities
  • Supports multiple imaging contrast methods: Brightfield (BF), Darkfield (DF), Phase Contrast (PH)
  • 1.4 MP monochrome CMOS camera with imaging software
  • On-board PC with Windows 10 and Accuva LEAP Software
  • 24 in. Full HD display
  • Motorized stage with trackball and stage inserts for various vessels Certifications and compliance
  • NATL Intertek ETL certified
  • RoHS and REACH compliant
  • UL and CSA approved
  • Designed and assembled in the USA
Specifications
Type Automated Imaging System
Camera 1.4 MP Monochrome CMOS
Voltage 110/220 V