Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ accuWash™ and accuWash Versa Microplate Washers

Catalog No. s28627 Shop All Fisherbrand Products
Change view
Click to view available options
:
accuWash Versa
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S28627 accuWash Versa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S28627

Includes

2 x 2L wash bottle, 1 x 2L rinse bottle, 1 x 4L waste bottle, 2 x 8 wash head, aerosol cover

Ease of use, convenience and versatility for washing microplates in ELISA applications

The Fisher Scientific accuWash and Fisher Scientific accuWash Versa are easy-to-use strip washers that offer convenience through a large color screen, intuitive graphical user interface, and a USB port. The accuWash is the basic model for washing 96-well plates only. It is intended for use when only a few similar assays are run routinely. The accuWash Versa can also wash cells and 384-well plates, offering enhanced flexibility needed for research use.

Certifications

CE mark

Warranty and Services

1 year

TRUSTED_SUSTAINABILITY

Specifications

Connections USB, RS-232-C
Wash/Waste Bottle Capacity Up to 3
Description accuWash Versa
Dispense Volume 50 to 400 μL (96); 20 to 120 μL (384)
Wash Heads 1 x 8, 1 x 12, 2 x 8, 2 x 12, and 1 x 16 wash heads; 2 x 8 cell wash head
Plate Type 96-well Plate, 384-well Plate
Residual Volume <2 μL
Weight (Metric) 9 kg
Wash Volume 50 to 1000 μL (96); 20 to 300 μL (384)
Display LCD Color Display, 4.3 in.
For Use With (Application) Immunoassays (ELISA); Cell Washing
For Use With (Equipment) Automation systems
Cell Washing Yes
Includes 2x2L wash bottle, 2 x 1L rinse bottle, 1x4L waste bottle, 2x8 wash head, aerosol cover
Dimensions (L x W x H) 15.2 x 13.6 x 9.4 in. (38.5 x 34.5 x 24 cm)
Weight (English) 20 lb.
Electrical Requirements 100/240 V 50/60 Hz
Voltage 100/240 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.