Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ ACE2, His tag (E. coli)

Catalog No. MP08720602 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1000 μg
250 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08720602 1000 μg
MP08720601 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP08720602 Supplier MP Biomedicals™ Supplier No. 08720602
Add to Cart
Add to Cart

Recombinant His-tagged Human ACE2 (Met1-Ser740) with polyhistidine tag at the N- and C-terminus. Expressed in E. coli.

  • The recombinant SARS-CoV-2 Nucleocapsid Protein consists of 740 amino acids and has a calculated molecular mass of 84.54 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 84.54 kDa
Name ACE2
Quantity 1000 μg
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His tag
Sterility Non-sterile
Reactivity Human
Tag Position N- and C-terminus
Expression System E. coli
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.