Learn More
AdipoGen ACE2 human ELISA Kit
SDP
Supplier:  AdipoGen AG45B0023KI01
Encompass_Preferred
Colorimetric Sandwich ELISA Assay. Sample Type Cell Culture Supernatant,Plasma,Serum,Urine. Range 0.0625 to 4ng/ml. Sensitivity 40pg/ml. Human ACE2 gene, localized in X chromosome, encodes for a 805 amino acid protein with an N-terminal signal sequence, a metalloprotease zinc binding site HEMGH and a hydrophobic region near the C-terminus acting as a membrane anchor transmembrane domain. Angiotensin-converting enzyme 2 ACE2 is a type I transmembrane metallocarboxypeptidase within the renin-angiotensin system RAS, which plays a key role in blood pressure regulation, fluid and electrolyte balance, thirst, cardiac/renal function and growth. ACE2 is expressed on the cell surface of type 2 alveolar epithelial cells in the lungs as well as on cells in many other tissues. ACE2 shares approximately 60% homology with ACE, the other key enzyme of the RAS system.
Catalog No. 50-256-2735
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.