Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACE2 Rabbit, anti-Human, Unconjugated, MP Biomedicals™

Catalog No. MP08L100004 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08L100004 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MP08L100004 Supplier MP Biomedicals, Inc Supplier No. 08L100004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ACE2 Rabbit Monoclonal Antibody has been experimentally validated in WB, IHC, IF assays.

  • The protein encoded by this gene belongs to the angiotensin-converting enzyme family of dipeptidyl carboxydipeptidases and has considerable homology to human angiotensin 1 converting enzyme
  • The organ- and cell-specific expression of this gene suggests that it may play a role in the regulation of cardiovascular and renal function, as well as fertility
  • Offers high specificity and potency against human ACE2 in immunological and biochemical assays and consistency between lots
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACE2
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Concentration 1.04 mg/mL
Conjugate Unconjugated
Dilution Western Blot: 1:500 to 1:2000; Immunohistochemistry: 1:50 to 1:200; Immunofluorescence: 1:50 to 1:200
Formulation In PBS buffer with 0.02% sodium azide, 0.05% BSA, 50% glycerol, pH7.3 to 7.4.
Gene Alias ACE-2; ACEH; ACE2
Host Species Rabbit
Immunogen Full-length human ACE2 protein
Purification Method Affinity Purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage -20°C
Form Liquid
Isotype IgG
Show More Show Less

For research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.