Learn More
Creative Biomart Recombinant Human ACE2 Protein, His-tagged
Supplier:  Creative Biomart ACE2359
Encompass_Preferred
The CHO derived ACE2 Human recombinant protein contains the extracellular domain amino acids 18-615 fused to Fc tag at C-terminal and has a molecular weight of ~130 kDa.
Catalog No. 50-208-4737
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.