Learn More
Creative Biomart Recombinant Human ACE2 Protein, His-tagged
Supplier:  Creative Biomart ACE2360
Encompass_Preferred
The HEK293 derived ACE2 Human recombinant protein contains the amino acids 18-740 fused to His tag at N-terminal. ACE2 Protein binds to SARS Coronavirus-2 [CoV-2019] Spike receptor binding domain.
Catalog No. 50-208-4738
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.