Learn More
ATCC ACE2plusC3-; A549/ACE2/TMPRSS2; Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL3560
Encompass
ACE2plusC3-A549/ACE2/TMPRSS2 is a cell line derived from the parental A549 cell line (ATCC CCL-185). This cell line can be used in antiviral screens, in SARS-CoV-2 biology and virus-host cell interaction studies, and in SARS-CoV-2 antibody neutralization assays.
Catalog No. 50-238-4765
Due to product restrictions, please to purchase or view availability for this product.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.