Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acemetacin 98.0+%, TCI America™
SDP

Catalog No. A245225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A245225G 25 g
A24525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A245225G Supplier TCI America Supplier No. A245225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53164-05-9
Molecular Formula C21H18ClNO6
Molecular Weight (g/mol) 415.826
MDL Number MFCD00151473
InChI Key FSQKKOOTNAMONP-UHFFFAOYSA-N
Synonym acemetacin, rantudil, emflex, acemetacinum, acemix, acemetacina, acemetacine, aximeixin, rheumibis, indomethacin carboxymethyl ester
PubChem CID 1981
ChEBI CHEBI:31162
IUPAC Name 2-[2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetyl]oxyacetic acid
SMILES CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)OCC(=O)O

Specifications

Melting Point 152°C
Quantity 25 g
UN Number 2811
Formula Weight 415.83
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Acemetacin
RTECSNumber NL3521400
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.