Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acenaphthene-d10 Standard, ISO 17034, 500 μg/mL, 1 mL

Catalog No. USATS1111
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USATS1111 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USATS1111 Supplier Agilent Technologies Supplier No. ATS1111
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15067-26-2 , 67-56-1
Molecular Formula C12H10
Molecular Weight (g/mol) 164.273
InChI Key CWRYPZZKDGJXCA-WHUVPORUSA-N
PubChem CID 177559
IUPAC Name 1,1,2,2,3,4,5,6,7,8-decadeuterioacenaphthylene
SMILES C1CC2=CC=CC3=C2C1=CC=C3

Specifications

CAS Max % 99.9, 0.06
Analyte or Component Names Acenaphthene-d10,methanol
Element Acenaphthene-d10
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 500μg/mL
Chemical Name or Material Acenaphthene-d10 Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.