Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acepromazine Maleate, USP, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18600166 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-166 5 g Amber Glass Bottle
18-600-167 25 g Amber Glass Bottle
18-600-163 100 g Amber Glass Bottle
18-600-165 500 g Amber Glass Bottle
18-600-168 1 kg Amber Glass Bottle
18-600-164 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-600-166 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A15715GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

A1571, 3598-37-6, C19H22N2OS·C4H4O4

Spectrum™ Chemical Acepromazine Maleate, USP is a phenothiazine derivative antipsychotic drug with veterinary sedative and antiemetic properties. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 3598-37-6
Molecular Formula C23H26N2O5S
Molecular Weight (g/mol) 442.53
InChI Key FQRHOOHLUYHMGG-WLHGVMLRSA-N
IUPAC Name (2E)-but-2-enedioic acid; 1-{10-[3-(dimethylamino)propyl]-10H-phenothiazin-2-yl}ethan-1-one
SMILES OC(=O)\C=C\C(O)=O.CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(C=C12)C(C)=O

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Melting Range 136-139°C
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 98 to 101%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.