Learn More
ACES, Biological Buffer, 98%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Specifications
Specifications
| CAS Max % | 100% |
| For Use With (Application) | Molecular Biology |
| Physical Form | Crystalline Powder |
| Quantity | 1 kg |
| Loss on Drying | 0.2% |
| Packaging | Amber Glass Bottle |
| Percent Purity | 98% |
| Grade | Reagent |
| Chemical Name or Material | N-(2-Acetamido)-2-Aminoethanesulfonic Acid |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.