Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetaldehyde ammonia trimer, 98%

Catalog No. AC148530500
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC148530500 50 g
AC148530250 25 g
AC148531000 100 g
AC148535000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC148530500 Supplier Thermo Scientific Chemicals Supplier No. 148530500

CAS: 58052-80-5 | C6H18N3 | 132.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58052-80-5
Molecular Formula C6H18N3
Molecular Weight (g/mol) 132.23
MDL Number MFCD00149559
InChI Key MZSSRMMSFLVKPK-UHFFFAOYSA-Q
Synonym 2,4,6-trimethyl-1,3,5-triazinane trihydrate, acetaldehyde ammonia trimer, hexahydro-2,4,6-trimethyl-1,3,5-triazine trihydrate, 1,3,5-triazine, hexahydro-2,4,6-trimethyl-, trihydrate, acetaldehyde ammonia trimer trihydrate, 2,4,6-trimethyl-1,3,5-triazaperhydroine, hydrate, hydrate, hydrate, acetaldehydeammoniatrimer, acmc-1bm0n, acetaldehyde-ammonia trimer trihydrate, acetaldehyde ammonia trimer nt
PubChem CID 2723814
SMILES CC1[NH2+]C(C)[NH2+]C(C)[NH2+]1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 50 g
Beilstein 26, I, 3
IUPAC Name 2,4,6-trimethyl-1,3,5-triazinane-1,3,5-triium
Formula Weight 183.26
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material Acetaldehyde ammonia trimer, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-868-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.