Learn More
Sigma Aldrich Fine Chemicals Biosciences Cas9 Neomycin Lenti Partic1EA
Supplier: Sigma Aldrich Fine Chemicals Biosciences LVCAS9NEO1EA
Ready-to-use Cas9 lentiviral particles are ideal for transducing a wide range of cell lines. The Cas9 Neomycin Lentiviral particles efficiently integrate Cas9 into the genomes of both dividing and non-dividing cell lines for use with CRISPR genome editing. Stably integrate Cas9 and Neo using the convenient ready-to-use lentiviral particles while avoiding the tedious lentivirus production process.