Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetaldehyde Semicarbazone 98.0+%, TCI America™
SDP

Catalog No. A019325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A019325G 25 g
A01935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A019325G Supplier TCI America Supplier No. A019325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 591-86-6
Molecular Formula C3H7N3O
Molecular Weight (g/mol) 101.109
MDL Number MFCD00059191
InChI Key IUNNCDSJWDQYPW-GORDUTHDSA-N
PubChem CID 5381348
IUPAC Name [(E)-ethylideneamino]urea
SMILES CC=NNC(=O)N

Specifications

Melting Point 164°C
Color White
Quantity 25 g
Formula Weight 101.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Acetaldehyde Semicarbazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.