Learn More
CPC Scientific Ac-Arg-Phe-Met-Trp-Met-Arg-NH2 (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific ACET002A
Encompass
Acetylated enkephalins, also named Opioid Receptor Antagonist, high affinity for m and K3 opioid receptor, weak affinity for K1 receptors and no affinity for K2 receptors.ONE-LETTER SEQUENCE: Ac-RFMWMR-NH2MOLECULAR FORMULA: C44H66N14O7S2MOLECULAR WEIGHT:967.23STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [152274-66-3], RESEARCH AREA: PainREFERENCES: C.T.Dooley et al., Proc. Natl. Acad. Sci. USA, 90, 10811 (1993)
Catalog No. 50-210-8584
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.