Learn More
Acetamide, 99%
Description
Acetamide is used as an industrial solvent, plasticizer, wetting and penetrating agent. It is also used for the transamidation of carboxamides in 1,4-dioxane. It finds application in lacquers and soldering flux. Further, it acts as a precursor to thioacetamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acetamide is used as an industrial solvent, plasticizer, wetting and penetrating agent. It is also used for the transamidation of carboxamides in 1,4-dioxane. It finds application in lacquers and soldering flux. Further, it acts as a precursor to thioacetamide.
Solubility
Soluble in water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
Specifications
Specifications
| Melting Point | 76°C to 81°C |
| Density | 1.159 |
| Boiling Point | 220°C to 222°C |
| Quantity | 5000 g |
| Beilstein | 1071207 |
| Sensitivity | Hygroscopic |
| Merck Index | 14,43 |
| Solubility Information | Soluble in water. |
| Formula Weight | 59.07 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
| Hazard Category | H351 |
| Hazard Statement | GHS H Statement H351 Suspected of causing cancer. |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| EINECSNumber | 200-473-5 |
| RTECSNumber | AB4025000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.