Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetamidoxime 97.0+%, TCI America™
SDP

Catalog No. A264625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A264625G 25 g
A26465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A264625G Supplier TCI America Supplier No. A264625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22059-22-9
Molecular Formula C2H6N2O
Molecular Weight (g/mol) 74.08
MDL Number MFCD00603514 MFCD00603514
InChI Key AEXITZJSLGALNH-UHFFFAOYSA-N
Synonym n'-hydroxyacetimidamide, n-hydroxyacetamidine, methylamidoxime, acetamide, oxime, 1z-n'-hydroxyethanimidamide, n-hydroxyethanimidamide, oximinoacetamide, e-n'-hydroxyacetimidamide, n'-hydroxyacetamidine, n-hydroxy-acetamidine
PubChem CID 5487681
IUPAC Name (Z)-N'-hydroxyethanimidamide
SMILES C\C(N)=N\O

Specifications

Melting Point 138°C
Color White
Quantity 25 g
Formula Weight 74.08
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Acetamidoxime
RTECSNumber AC6900000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.