Learn More
Acetanilide, 98%
Description
Acetanilide is used as an intermediate in the synthesis of rubber accelerator, dyes and camphor. It is also used in the synthesis of penicillin and other pharmaceutical products. It is involved in the preparation of 4-acetamidobenzenesulfonyl chloride, which is an intermediate during the synthesis of sulfa drugs. Further, it is employed as a experimental photographic developer. In addition to this, it is used to stabilize cellulose ester varnishes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acetanilide is used as an intermediate in the synthesis of rubber accelerator, dyes and camphor. It is also used in the synthesis of penicillin and other pharmaceutical products. It is involved in the preparation of 4-acetamidobenzenesulfonyl chloride, which is an intermediate during the synthesis of sulfa drugs. Further, it is employed as a experimental photographic developer. In addition to this, it is used to stabilize cellulose ester varnishes.
Solubility
Soluble in ethanol, diethyl ether, acetone, benzene. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
| Melting Point | 113°C to 116°C |
| Density | 1.121 |
| Boiling Point | 304°C to 305°C |
| Flash Point | 161°C (321°F) |
| Assay Percent Range | 98% |
| Quantity | 2500 g |
| Beilstein | 606468 |
| Merck Index | 14,50 |
| Solubility Information | Soluble in ethanol,diethyl ether,acetone,benzene. Slightly soluble in water. |
| Formula Weight | 135.17 |
| Show More |
Safety and Handling
| Hazard Category | H302 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| EINECSNumber | 203-150-7 |
| RTECSNumber | AD7350000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.