Learn More
Cytiva Acetate 5.0, 50mL
Catalog No. 501055295 Shop All Cytiva Products
Encompass
Immobilization buffer, 10 mM sodium acetate pH 5.0
Catalog No. 50-105-5295 Supplier Cytiva Supplier No. BR100351
Immobilization buffer, 10 mM sodium acetate pH 5.0