Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetate Buffer, for Iron (Phenanthroline Method), Ricca Chemical
SDP

Catalog No. 6016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6016 500 mL Natural Poly Bottle
60-32 1 L Natural Poly Bottle
601 4 L Natural Poly Bottle
R006000020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6016 Supplier Ricca Chemical Company Supplier No. 6016
Add to Cart
Add to Cart

For Analysis of Iron (pHenanthroline Method)

Specifications

Chemical Name or Material Acetate Buffer
CAS 64-19-7, 631-61-8, 7732-18-5
CAS Min % 63.0, 21.6, 12.95
Color Colorless
CAS Max % 66.0, 22.48, 13.48
Concentration 1X
For Use With (Application) Iron Analysis
Physical Form Liquid
Quantity 500 mL
Solution Type Buffer
Packaging Natural Poly Bottle
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.