Learn More
Acetate Buffer, pH 3.5 For USP Heavy Metals Test, Reagents
Specifications
Specifications
| Chemical Name or Material | Acetate Buffer, pH 3.5 For USP Heavy Metals Test |
| CAS | 7732-18-5, 631-61-8, 631-61-8 |
| Boiling Point | 118°C |
| Color | Colorless |
| pH | 3.5 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Solution Type | Buffer |
| Packaging | Poly Bottle |
| Specific Gravity | 1.0737 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.