Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, 25% (v/v), Ricca Chemical
SDP

Catalog No. 1382516 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Hard Pack
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
13825-16 500 mL Natural Poly Bottle
13825-32 1 L Natural Poly Bottle
13825-1 4 L Natural Poly Bottle
138255 20 L Cubitainer
13-622-600 20 L Hard Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 13825-16 Supplier Ricca Chemical Company Supplier No. 138.2516
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) Mixture
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Min % 74.0, 24.0
CAS Max % 76.0, 26.0
Percent Purity 25% v/v
Grade Laboratory
Concentration or Composition (by Analyte or Components) 25% (v/v) Aqueous Solution
Quantity 500 mL
Packaging Natural Poly Bottle
Chemical Name or Material Acetic Acid, 25% (v/v)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.