Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, 30% (v/v), Ricca Chemical
SDP

Catalog No. 1383032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
1383032 1 L Natural Poly Bottle
138301 4 L Natural Poly Bottle
1383025 10 L Cubitainer
138305 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 1383032 Supplier Ricca Chemical Company Supplier No. 138.3032
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 7732-18-5 , 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) Mixture
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Min % 75.0, 23.48
Color Colorless
CAS Max % 78.0, 24.43
Concentration 30% (v/v)
Percent Purity 30% v/v
Grade Laboratory
Quantity 1 L
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.