Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid-d{4}, 99.5% (Isotopic)

Catalog No. AA3646906
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3646906 5 g
AA3646909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3646906 Supplier Thermo Scientific Chemicals Supplier No. 03646906
Add to Cart

CAS: 1186-52-3 | C2H4O2 | 64.08 g/mol

Acetic acid-d{4} is used as a deuterated, polar and protic solvent, in food preservatives, PH control agent, flavor enhancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1186-52-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 64.08
MDL Number MFCD00051051
InChI Key QTBSBXVTEAMEQO-GUEYOVJQSA-N
Synonym acetic acid-d4, acetic-d3 acid-d, 2h3 acetic 2h acid, tetradeuteroacetic acid, 2 h? ethan 2 h oic acid, acetic acid d4, 2h4 acetic acid, cd3cood, acetic acid d, acetic acid-d4, 99.5 atom % d
PubChem CID 2723903
SMILES [2H]OC(=O)C([2H])([2H])[2H]

Specifications

Physical Form Liquid
Quantity 5 g
Sensitivity Moisture
Flash Point 40°C (104°F)
Density 1.12 g/mL
Odor Pungent, makes eyes water
Melting Point 17°C
Boiling Point 118°C
UN Number UN2789
Solubility Information Soluble in water,ethanol and ether.
IUPAC Name deuterio 2,2,2-trideuterioacetate
Formula Weight 64.08
Percent Purity 99.5%
Chemical Name or Material Acetic acid-d4, Isotopic
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETIC ACID, GLACIAL
EINECSNumber 214-693-4
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.