accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1186-52-3

IUPAC Name: (²H₃)ethan(²H)oic acid
Synonyms: (²H₃)ethan(²H)oic acid
Molecular Weight (g/mol): 64.08
Molecular Formula: C2H4O2
InChi Key: QTBSBXVTEAMEQO-GUEYOVJQSA-N
SMILES: [2H]OC(=O)C([2H])([2H])[2H]
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results